Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asoxime dichloride | 34433-31-3 | MFCD00869535 | ≥98.0% | 359.2 g/mol | C14H16Cl2N4O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY106901A10MG

Encompass_Preferred

Asoxime dichloride is an oxime-class cholinesterase reactivator used in research to reverse organophosphate-induced acetylcholinesterase inhibition. Supplied as the dichloride salt, it is applied in biochemical and pharmacological studies of nerve agent antidotes and cholinergic signaling.

  • High purity, typically ≥98%.
  • Dichloride salt form for stability and solubility.
  • Used as a research tool in studies of organophosphate poisoning and antidote development.
  • Molecular weight 359.2 g/mol; formula C14H16Cl2N4O3.
  • Available in small research quantities, suitable for in vitro and in vivo studies.

Catalog No. 50-002-39888


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.