Learn More
Medchemexpress LLC ASP-4058 | 952565-91-2 | 99.54% | 442.31 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY11102110MM/1ML
Encompass_Preferred
ASP-4058 is a next-generation, selective and oral bioactive agonist for Sphingosine 1-Phosphate receptors 1 and 5 (S1P1 and S1P5). It ameliorates rodent experimental autoimmune encephalomyelitis with a favorable safety profile. It is for research use only and not sold to patients.
  • Next-generation, selective, and oral bioactive agonist for S1P1 and S1P5 receptors.
  • Ameliorates rodent experimental autoimmune encephalomyelitis.
  • Favorable safety profile.
  • Reduces clinical score in a dose-dependent manner in EAE rats.
  • Prevents decreases in body weight of EAE rats.
  • Maintains clinical score at a relatively low level in EAE mice.
Catalog No. 50-003-43349