Learn More
Medchemexpress LLC ASP-8731 | 2488255-42-9 | 98.4% | 395.41 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16787410MM/1ML
Encompass_Preferred
ASP-8731 is an orally active BACH1 inhibitor that activates antioxidant, anti-inflammatory, and globin gene pathways by relieving BACH1's inhibitory effect on NRF2-mediated gene transcription. It upregulates HMOX1, FTH1, and various globins (HGB, HBG, HBA), increasing fetal hemoglobin (HbF) and inducing F-cell production. It also reduces inflammatory responses, white blood cell counts, heme-induced glutathione depletion, and microcirculatory stasis, showing potential for inhibiting sickle cell disease and related hematological disorders.
  • Orally active BACH1 inhibitor
  • Activates antioxidant, anti-inflammatory, and globin gene pathways
  • Increases fetal hemoglobin (HbF) levels
  • Reduces inflammation and white blood cell counts
  • Blocks heme-induced glutathione depletion and microcirculatory stasis
  • Potential for sickle cell disease treatment
Catalog No. 50-003-93575