Learn More
Medchemexpress LLC ASP-9521 | 1126084-37-4 | 98.0% | 330.42 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19903100MG
Encompass_Preferred
ASP-9521 is a potent, selective, and orally available AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3, intended for research use only.
  • Potent, selective, and orally available AKR1C3 inhibitor.
  • IC50 of 11 nM for human AKR1C3 and 49 nM for monkey AKR1C3.
  • Shows more than 100-fold selectivity for AKR1C3 over the isoform AKR1C2.
  • Suppresses AD-dependent PSA production and cell proliferation in cells.
  • Inhibits AD-induced intratumoural T production in xenografts.
  • Bioavailability after oral administration is 35% in rats, 78% in dogs, and 58% in monkeys.
Catalog No. 50-004-01740