Learn More
Medchemexpress LLC ASP-9521 | 1126084-37-4 | 98.0% | 330.42 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY19903200MG
Encompass_Preferred
ASP-9521 is a potent, selective, and orally available AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3. It shows more than 100-fold selectivity for AKR1C3 over the isoform AKR1C2. This inhibitor suppresses AD-dependent PSA production and cell proliferation in LNCaP-AKR1C3 cells, and inhibits AD-induced intratumoural T production in CWR22R xenografts for up to 24 hours.
  • Potent and selective AKR1C3 inhibitor
  • Orally available
  • High selectivity over AKR1C2
  • Suppresses AD-dependent PSA production and cell proliferation
  • Inhibits AD-induced intratumoural T production
  • Inhibitory effect maintained for 24 hours
Catalog No. 50-004-01739