Learn More
Medchemexpress LLC Asp-9521 | 1126084-37-4 | 98.0% | 330.42 | 50 MG

Supplier: Medchemexpress LLC HY1990350MG
ASP-9521 is a potent, selective and orally available AKR1C3 inhibitor with an IC50 of 11 nM for human AKR1C3.
- IC50: 11 nM (human AKR1C3), 49 nM (monkey AKR1C3)
- Inhibits conversion of androstenedione (AD) into androstenediol and testosterone (T) by recombinant human or cynomolgus AKR1C3.
- Shows more than 100-fold selectivity for AKR1C3 over the isoform AKR1C2.
- Suppresses AD-dependent PSA production and cell proliferation in LNCaP-AKR1C3 cells.
- Single oral administration (3 mg/kg) inhibits AD-induced intratumoural T production in CWR22R xenografts, with the inhibitory effect maintained for 24 h.
- Rapidly eliminated from plasma after oral administration, while intratumoural concentration remained high.
- Bioavailability after oral administration (1 mg/kg) is 35% in rats, 78% in dogs, and 58% in monkeys.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.