Learn More
Medchemexpress LLC Asp-Asp-Asp-Asp-Asp-Asp | 145224-96-0 | 99.5% | 708.54 | 10 MG

Supplier: Medchemexpress LLC HYP382310MG
Asp-Asp-Asp-Asp-Asp-Asp is a polyaspartic acid used in research related to influenza virus neuraminidase. It is a solid, white to off-white substance with high purity.
- High purity of 99.5%
- Molecular weight of 708.54
- Supports research on influenza virus neuraminidase
- Requires sealed storage, away from moisture and light, under nitrogen
- Powder form stores for 2 years at -80°C or 1 year at -20°C
- Solution form stores for 6 months at -80°C or 1 month at -20°C
- Can be shipped at room temperature if transit time is less than 2 weeks
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.