Learn More
Medchemexpress LLC ASP4132 | 1640294-30-9 | 99.8% | 937.06 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13644710MM/1ML
Encompass_Preferred
ASP4132 is an orally active, potent AMPK activator with an EC50 of 18 nM. It exhibits anti-cancer activity and leads to tumor regression in breast cancer xenograft mouse models.
  • Potent AMPK activator with an EC50 of 18 nM
  • Exhibits anti-cancer activity
  • Leads to tumor regression in breast cancer xenograft mouse models
  • Shows cell growth inhibitory activity against MDA-MB-453 breast cancer cells (IC50 = 0.014 μM)
  • Stable in human liver microsomes
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-003-50311