Learn More
Medchemexpress LLC Asp4132 | 1640294-30-9 | 99.8% | 937.06 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136447100MG
Encompass_Preferred
ASP4132 is an orally active, potent AMPK activator with an EC50 of 18 nM. It demonstrates anti-cancer activity and induces tumor regression in breast cancer xenograft mouse models. It is for research use only.
  • Potent AMPK activator
  • Exhibits anti-cancer activity
  • Induces tumor regression in breast cancer models
  • Orally active compound
  • Suitable for in vitro and in vivo studies
Catalog No. 50-003-48530