Learn More
Medchemexpress LLC ASP4132 | 1640294-30-9 | 99.8% | 937.06 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY136447
Encompass_Preferred
ASP4132 is an orally active, potent AMPK activator with an EC50 of 18 nM. It exhibits anti-cancer activity and induces tumor regression in breast cancer xenograft mouse models.
  • Orally active, potent AMPK activator.
  • Has anti-cancer activity.
  • Causes tumor regression in breast cancer xenograft mouse models.
  • Shows comparable cell growth inhibitory (IC50=0.014 μM) activity against MDA-MB-453 breast cancer cells.
  • Stable in human liver microsomes.
Catalog No. 50-003-51423