Learn More
Medchemexpress LLC ASP4132 | 1640294-30-9 | 99.8% | 937.06 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13644750MG
Encompass_Preferred
ASP4132 is an orally active, potent AMPK activator with an EC50 of 18 nM. It demonstrates significant anti-cancer activity and induces tumor regression in breast cancer xenograft mouse models.
  • Potent AMPK activator with an EC50 of 18 nM.
  • Exhibits anti-cancer activity.
  • Induces tumor regression in breast cancer xenograft models.
  • Shows cell growth inhibitory activity against MDA-MB-453 breast cancer cells.
  • Stable in human liver microsomes.
Catalog No. 50-003-49668