Learn More
Medchemexpress LLC ASP4132 | 1640294-30-9 | 99.8% | 937.06 | C46H51F3N6O8S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1364475MG
Encompass_Preferred
ASP4132 is an orally active small-molecule activator of AMP-activated protein kinase (AMPK) that shows potent activity (EC50 = 18 nM) and reported antitumor effects in preclinical models, including tumor regression in breast cancer xenograft studies. It is supplied at high purity for laboratory research use.
  • Potent AMPK activator (EC50 = 18 nM).
  • Orally bioavailable with demonstrated in vivo efficacy.
  • Reported antitumor activity in breast cancer xenograft models.
  • High purity (~99.8%) suitable for research applications.
  • Available in multiple small-scale sizes and solution formulations.
Catalog No. 50-002-21879