Learn More
Selleck Chemical LLC ASP4132 S8953-25MG
SDP
Supplier:  Selleck Chemical LLC S895325MG
Encompass_Preferred
ASP4132 is a potent and orally active activator of Adenosine Monophosphate-Activated Protein Kinase (AMPK) with EC50 of 0 018 M ASP4132 is used as a clinical candidate for the treatment of human cancer
Catalog No. 50-000-24080