Learn More
Lkt Laboratories CAS 9015-68-3, ≥98%, Catalyzes hydrolysis of asparagine to aspartate and ammonia. It treats leukemias by depleting asparagine levels and inhibiting cell growth.
SDP
Supplier:  Lkt Laboratories A7460
Encompass_Preferred
Asparaginase is an enzyme that catalyzes the hydrolysis of asparagine to aspartate and ammonia, depleting asparagine levels and inhibiting cell growth; it is commonly used as an anticancer chemotherapeutic and anti-angiogenic compound in the treatment of leukemia. Asparaginase inhibits proliferation of acute myelogenous leukemia (AML) cells, inducing apoptosis mediated by apoptosis-inducing factor (AIF). Asparaginase decreases β1 integrin binding and phosphorylation of FAK, inducing autophagy; this decreases microvascular endothelial cell tube formation and inhibits invasion of ovarian cancer cells. In clinical settings, asparaginase may induce thrombotic effect, as administration often results in depletion of anti-thrombin, and it may also increase serum triglycerides.
Catalog No. NC2539814