Learn More
CPC Scientific H-Asp-Phe-OMe (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ASPA001A
Encompass
This dipeptide is 180 to 200 times sweeter than that of sucrose.ONE-LETTER SEQUENCE: DF-OMeMOLECULAR FORMULA: C14H18N2O5MOLECULAR WEIGHT:294.31STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [22839-47-0], RESEARCH AREA: ObesityREFERENCES: Xia Li, et al. Chem Senses, Jun 2011; 36: 453-475.
Catalog No. 50-210-8789