Learn More
Medchemexpress LLC Aspartic-2,3,3-d3 acid | 14341-75-4 | 136.12 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0666S2
Encompass_Preferred
DL-Aspartic acid-d3 is a deuterium-labeled form of L-Aspartic acid, an amino acid known for its ability to penetrate the blood-brain barrier. It is utilized in the preparation of prodrugs for targeted delivery to colon and cecal tissues and in studies related to inflammatory conditions. This compound serves as a valuable tool for research applications, particularly as a tracer and an internal standard in various quantitative analytical techniques.
  • Used for preparing prodrugs for targeted delivery to colon and cecal tissues.
  • Applied in the study of inflammatory conditions.
  • Functions as a tracer in research.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Features deuterium labeling, which can affect pharmacokinetic and metabolic profiles.
Catalog No. 50-003-89560