Learn More
Medchemexpress LLC (-)-Aspartic acid | 1783-96-6 | 98.0% | 133.10 | 100 G
SDP
Supplier:  Medchemexpress LLC HY42068100G
Encompass_Preferred
(-)-Aspartic acid is a pyroptosis inhibitor, neurotransmitter, and neuromodulator. It plays a crucial role in hormone synthesis and regulation, as well as the development of the nervous and endocrine systems. This product is intended for research use only.
  • Inhibits HGF cell pyroptosis to prevent gingival fibroblast inflammation.
  • Acts as a neurotransmitter and neuromodulator.
  • Participates in hormone synthesis and regulation.
  • Plays a role in nervous system and endocrine system development.
  • Stimulates steroid production in mammalian Leydig cell lines.
  • Acts as a neuropeptide co-transmitter.
  • Activates cAMP dependent gene transcription and inhibits CREB function.
  • Reduces BDNF expression and induces excitotoxic neuronal death.
  • Participates in testosterone synthesis.
  • Increases secretion of GnRH, PRL, LH, and GH.
Catalog No. 50-004-04394