Learn More
Medchemexpress LLC (-)-Aspartic acid | 1783-96-6 | 98.0% | 133.10 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY42068
Encompass_Preferred
(-)-Aspartic acid is a pyroptosis inhibitor that functions as a neurotransmitter and neuromodulator. It is involved in hormone synthesis and regulation, nervous system development, and the endocrine system. This product is for research use only.
  • Inhibits pyroptosis
  • Acts as a neurotransmitter and neuromodulator
  • Participates in hormone synthesis and regulation
  • Plays a role in nervous system development and the endocrine system
  • Prevents gingival fibroblast inflammation
  • Stimulates steroid production in mammalian Leydig cell lines
  • Acts as a neuropeptide co-transmitter by releasing glutamate or GABA
  • Participates in testosterone synthesis
  • Increases secretion of various hormones like GnRH, PRL, LH, and GH
Catalog No. 50-004-02878