Learn More
Medchemexpress LLC Aspartic acid calcium | 10389-09-0 | 171.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN0666B10MM/1ML
Encompass_Preferred
Aspartic acid calcium (Calcium L-aspartate) is a chelate where calcium is attached to L-Aspartic acid. It is an amino acid that can penetrate the blood-brain barrier and is commonly used for preparing prodrugs to target colon and cecal tissues. This L-isomer of aspartic acid is also utilized in the study of inflammatory conditions and is one of twenty-three proteinogenic amino acids directly incorporated into proteins.
  • Can penetrate the blood-brain barrier.
  • Commonly used for preparing prodrugs to target colon and cecal tissues.
  • Used in the study of inflammatory conditions.
  • Directly incorporated into proteins as one of twenty-three proteinogenic amino acids.
Catalog No. 50-003-88380