Learn More
ATCC Aspergillus brasiliensis; WLRI 034(120); 6 x 0.3mL
Supplier:  ATCC 16404MINIPACK
Encompass
ATCC 16404-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 16404™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-2515