Learn More
Medchemexpress LLC Asperuloside | 14259-45-1 | MFCD31725921 | 414.36 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN138220MG
Encompass_Preferred
Asperuloside is an iridoid isolated from Hedyotis diffusa, with anti-inflammatory activity. It inhibits inducible nitric oxide synthase (iNOS) and suppresses NF-κB and MAPK signaling pathways.
  • Anti-inflammatory activity.
  • Inhibits inducible nitric oxide synthase (iNOS).
  • Suppresses NF-κB and MAPK signaling pathways.
  • Inhibits productions of NO, PGE2, TNF-α and IL-6 in LPS-induced RAW 264.7 cells (In Vitro).
  • Inhibits cell viability and induces ER stress-dependent apoptosis in leukemia cell lines (U937, HL-60, AML) (In Vitro).
  • Reduces food intake, body weight, and adipose masses in rats consuming a high fat diet (HFD) (In Vivo).
  • Inhibits tumor growth of U937 xenografts mice model (In Vivo).
  • Relieves LPS-induced acute lung injury via inhibiting MAPK and NF-κB signaling in BALB/c mice (In Vivo).
Catalog No. 50-004-16269