Learn More
Medchemexpress LLC HY-N1382 5mg Medchemexpress, Asperuloside CAS:14259-45-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN13825MG
Encompass_Preferred
Medchemexpress, HY-N1382 5mg Asperuloside CAS:14259-45-1 Asperuloside is an iridoid isolated from Hedyotis diffusa, with anti-inflammatory activity. Asperuloside inhibits inducible nitric oxide synthase (iNOS), suppresses NF-κB and MAPK signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2840