Learn More
Aviva Systems Biology Corp ASPH Antibody - N-terminal region (ARP38449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASPH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASPH. Alternate names: BAH, CASQ2BP1, HAAH, JCTN, junctin, AAHApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-014-56661