Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aspirin Aluminum | 23413-80-1 | 99.0% | 5 G
SDP

Supplier:  Medchemexpress LLC HYB21755G

Encompass_Preferred

Aspirin Aluminum is a novel intermolecular compound that can inhibit gastrointestinal mucosal disorders induced by NSAIDs (non-steroidal anti-inflammatory agents).

  • Can inhibit gastrointestinal mucosal disorders induced by NSAIDs
  • Molecular weight: 402.29
  • Formula: C18H15AlO9
  • Appearance: Solid
  • Color: White to off-white
  • Stable under recommended storage conditions
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-10968


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.