Learn More
Medchemexpress LLC Aspirin-d3 (Acetylsalicylic acid-d3) | 921943-73-9 | 99.8% | 183.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14654S100MG
Encompass_Preferred
Aspirin-d3 (Acetylsalicylic acid-d3; ASA-d3) is a deuterium-labeled Aspirin and an orally active, potent, and irreversible inhibitor of cyclooxygenase COX-1 and COX-2. It induces apoptosis, inhibits NF-κB activation, and inhibits platelet prostaglandin synthetase, which can prevent coronary artery and cerebrovascular thrombosis. Used for research, its stable heavy isotopes serve as tracers for quantitation, and deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Aspirin
  • Orally active, potent, and irreversible inhibitor of cyclooxygenase COX-1 and COX-2
  • Induces apoptosis
  • Inhibits activation of NF-κB
  • Inhibits platelet prostaglandin synthetase
  • Can prevent coronary artery and cerebrovascular thrombosis
  • Utilizes stable heavy isotopes as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-63048