Learn More
Hach Company pH Chemkey Reagents (box of 25)
Supplier:  Hach Company 9759000
Encompass
pH Chemkeys, range 6.3 - 9.0. For SL1000 Portable Parallel Analyzer (PPA).Avoid the hassle of pH probes and calibration in the field with our easy to use colorimetric pH Chemkey. The Hach SL1000 Portable Parallel Analyzer (PPA) performs the same tests with less than half the manual steps. Get highly accurate results, with less opportunity for errors, in a fraction of the time. Up to six parameters, tested in parallel. The SL1000 is a portable colorimeter that provides faster testing by using Chemkey reagents instead of powder pillows. All chemicals and processes are entirely contained inside the Chemkey.
Catalog No. 50-199-7369