Learn More
Hach Company High Range Total Alkalinity Chemkey Reagents (box of 25)
Supplier:  Hach Company 8636100
Encompass
High Range Total Alkalinity Chemkeys, range 200 - 700 mg/L CaCO3. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.
Catalog No. 50-199-7272