Learn More
Hach Company Pump Tube Adapter, Large Collar - Replacement for Lachat FIA Pump Assemblies
Supplier:  Hach Company 85293
Encompass
Pump tube adapter with a large collar designed as a replacement component for flow-injection analysis (FIA) pump assemblies. It secures pump tubing in the pump head to maintain alignment and reduce leakage during sampling and analysis. Detailed material and dimension specifications are not provided on the public product page.
  • Provides a secure connection for pump tubing.
  • Large collar improves retention and alignment within the pump head.
  • Suitable for flow-injection analysis pump assemblies.
  • Easy to install as a direct replacement during routine maintenance.
  • Verify tubing compatibility and fit before installation.
Catalog No. NC2017462