Learn More
Selleck Chemical LLC Astaxanthin 5mg 472-61-7 -Carotene-4,4'-dione, Trans-Astaxanthin
SDP
Supplier:  Selleck Chemical LLC S38345MG
Encompass_Preferred
Astaxanthin (?-Carotene-4,4'-dione, Trans-Astaxanthin), a xanthophyll carotenoid, is a nutrient with unique cell membrane actions and diverse clinical benefits with excellent safety and tolerability. Astaxanthin, a red dietary carotenoid isolated from Haematococcus pluvialis, is a modulator of PPAR? and a potent antioxidant with antiproliferative, neuroprotective and anti-inflammatory activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8323