Learn More
Thermco Products Inc ASTMCERT THERM 150,165,180,200
Supplier:  Thermco Products Inc ACC103CFC
Encompass_Preferred
ASTM 103C thermometer (Hg) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC1239951