Learn More
Selleck Chemical LLC Astragaloside A S2415-50mg
SDP
Supplier:  Selleck Chemical LLC S241550MG
Encompass_Preferred
Astragaloside A (Astragaloside IV) is the primary pure saponin isolated from Astragalus membranaceus which has been widely used for the treatment of cardiovascular diseases
Catalog No. 50-311-2971