Learn More
TARGETMOL CHEMICALS INC ASTRAGALOSIDE II 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T392410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Astragaloside II (Astrasieversianin VIII) is an effective MDR reversal agent and may be a potential adjunctive agent for hepatic cancer chemotherapy. Astragaloside II also induces osteogenic activities differentiation proliferation and mineralization through the bone morphogenetic protein-2/MAPK and Smad1/5/8 pathways. purity: 99%
Catalog No. 50-257-6858