Learn More
Selleck Chemical LLC Astragaloside IV S3901-100mg
SDP
Supplier:  Selleck Chemical LLC S3901100MG
Encompass_Preferred
Astragaloside IV (AST-IV AS-IV) is a bioactive saponin first isolated from the dried plant roots of the genus Astragalus which is used in traditional Chinese medicine It has various effect on the cardiovascular immune digestive and nervous systems AS-IV suppresses activation of p-Akt p-mTOR p-NF- B and p-Erk1/2
Catalog No. 50-311-0398