Learn More
Medchemexpress LLC Astragenol | 86541-79-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN792410MG
Encompass_Preferred
Astragenol is an intermediate used for the synthesis of its derivatives. These derivatives have shown promise as anti-inflammatory agents, particularly in prostate cancer research.
  • Intermediate for derivative synthesis
  • Derivatives act as anti-inflammatory agents
  • Relevant for prostate cancer research
Catalog No. 50-004-17210