Learn More
Medchemexpress LLC Astragenol | 86541-79-9 | 100 MG

Supplier: Medchemexpress LLC HYN7924100MG
Astragenol is an intermediate employed in the synthesis of Astragenol derivatives. These derivatives show potential as anti-inflammatory agents and are being investigated for their role in prostate cancer research.
- Purity: 98.0%
- Molecular weight: 490.72
- Formula: C30H50O5
- Appearance: Solid
- Color: White to off-white
- Structure classification: Terpenoids, Triterpenes
- Initial source: Plants, other families
- For research use only
- Storage: -20°C, sealed, away from moisture. In solvent: -80°C (6 months), -20°C (1 month)
- In vitro solubility: Ethanol: 50 mg/mL (101.89 mM; needs ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.