Learn More
Medchemexpress LLC Astragenol | 86541-79-9 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN7924
Astragenol is an intermediate utilized for the synthesis of Astragenol derivatives. Astragenol derivatives are recognized as promising anti-inflammatory agents for prostate cancer research.
- Purity: 98.0%
- Molecular weight: 490.72
- Formula: C30H50O5
- Appearance: Solid
- Color: White to off-white
- Structure classification: Terpenoids, triterpenes
- Initial source: Plants other families
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.