Learn More
Medchemexpress LLC Astragenol | 86541-79-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN79245MG
Encompass_Preferred
Astragenol is an intermediate primarily utilized for the synthesis of its derivatives. These derivatives have shown promise as anti-inflammatory agents in prostate cancer research.
  • Intermediate for derivative synthesis
  • Potential for anti-inflammatory research
  • High purity (98.0%)
  • Solid, white to off-white appearance
  • Classified as a triterpene
  • Plant-derived
  • Soluble in ethanol
  • Stable with recommended storage
Catalog No. 50-004-19385