Learn More
Medchemexpress LLC Astragenol | 86541-79-9 | 50 MG

Supplier: Medchemexpress LLC HYN792450MG
Astragenol is an intermediate used for Astragenol derivative synthesis. Astragenol derivatives are promising anti-inflammatory agents for prostate cancer research.
- Molecular weight: 490.72
- Molecular formula: C30H50O5
- Appearance: Solid
- Color: White to off-white
- Purity: 98.0%
- Store at -20°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture).
- Soluble in ethanol at 50 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.