Learn More
Medchemexpress LLC HY-P0257 1mg Medchemexpress, Astressin CAS:170809-51-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02571MG
Encompass_Preferred
Medchemexpress, HY-P0257 1mg Astressin CAS:170809-51-5 Astressin is a potent corticotropin releasing factor ( CRF ) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4082