Learn More
Medchemexpress LLC Astringin (trans-Astringin) | 29884-49-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN40935MG
Encompass_Preferred
Astringin (trans-Astringin) is an orally active natural flavonoid compound with multiple activities including anti-oxidation, anti-inflammation, anti-apoptosis, and ferroptosis inhibition. It is utilized in research for diseases such as acute lung injury, where it inhibits the production of oxidative stress and inflammatory factors.
  • Orally active natural flavonoid compound
  • Exhibits anti-oxidation, anti-inflammation, and anti-apoptosis activities
  • Functions as an inhibitor of ferroptosis
  • Suppresses the elevation of reactive oxygen species (ROS) and malondialdehyde (MDA)
  • Reduces inflammatory factors like TNF-α, IL-1β, and IL-6
  • Inhibits PI3K/AKT/NF-κB and TLR4/MyD88/HMGB1/RAGE pathways
  • Enhances cell viability and inhibits apoptosis in vitro
  • Alleviates chromium-induced oxidative stress, inflammation, and renal tissue damage in vivo
  • Improves renal function and restores biochemical parameters
Catalog No. 50-004-19744