Learn More
Medchemexpress LLC Astringin (trans-astringin) | 29884-49-9 | 99.0% | 406.38 g/mol | C20H22O9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN409310MG
Encompass_Preferred
Astringin (trans-astringin) is a natural flavonoid compound used as a research standard for studies of oxidative stress, inflammation, and related pathways. It is supplied as a solid analytical standard intended for in vitro and in vivo research applications.
  • High chemical purity (≈99%) suitable for analytical work.
  • Light yellow to yellow solid appearance for easy visual inspection.
  • Available in small research quantities (e.g., 10 mg) for assay and screening workflows.
  • Stable when stored as a powder at -20°C for long-term storage.
  • Reported bioactivity includes antioxidant, anti-inflammatory, and anti-apoptotic effects for use in pathway studies.
Catalog No. 50-002-89806