Learn More
TARGETMOL CHEMICALS INC ASTX660 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ00295MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ASTX660 is an orally bioavailable dual antagonist of cIAP and XIAP. purity: 99%
Catalog No. 50-257-8845