Learn More
Medchemexpress LLC HY-109565 5mg Medchemexpress, ASTX660 CAS:1799328-86-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1095655MG
Encompass_Preferred
Medchemexpress, HY-109565 5mg ASTX660 CAS:1799328-86-1 ASTX660 is an orally bioavailable dual antagonist of cellular inhibitor of apoptosis protein (cIAP) and X-linked inhibitor of apoptosis protein (XIAP). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0771