Learn More
Cayman Chemical AsukamycIn 2.5mg
SDP
Supplier:  Cayman Chemical 201332.5 MG
Encompass_Preferred
An antibiotic that inhibits growth in various tumor cell lines (IC50s = 1-5 µM) by activating caspases 8 and 3
Catalog No. 50-298-1265