Learn More
Cayman Chemical AsunaprevIr 10mg
SDP
Supplier:  Cayman Chemical 2083510 MG
Encompass_Preferred
An inhibitor of HCV NS3/4A serine protease; a direct-acting inhibitor of hepatitis C virus (HCV)
Catalog No. 50-298-3807