Learn More
Medchemexpress LLC HY-14434 10mg Medchemexpress, Asunaprevir CAS:630420-16-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1443410MG
Encompass_Preferred
Medchemexpress, HY-14434 10mg Asunaprevir CAS:630420-16-5 Asunaprevir (BMS-650032) is a potent and orally bioavailable hepatitis C virus (HCV) NS3 protease inhibitor, with IC50 of 0.2 nM-3.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9050