Learn More
TARGETMOL CHEMICALS INC AT-121 hydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T395291MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. AT-121 hydrochloride is a dual agonist of nociception and mu opioid receptor with Ki values of 3.67 and 16.49 nM, respectively.AT-121 hydrochloride is a safe, non-addictive, pain relieving compound with anti-injury and analgesic activity. AT-121 hydrochloride is a safe, non-addictive, pain relieving compound with anti-injury and analgesic activity. Purity 97.28%
Catalog No. 50-287-9173