Learn More
Selleck Chemical LLC AT13148 5mg 1056901-62-2
SDP
Supplier:  Selleck Chemical LLC S75635MG
Encompass_Preferred
AT13148 is an oral, ATP-competitive, multi-AGC kinase inhibitor with IC50 of 38 nM/402 nM/50 nM, 8 nM, 3 nM, and 6 nM/4 nM for Akt1/2/3, p70S6K, PKA, and ROCKI/II, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0959775