Learn More
Medchemexpress LLC At-533 | 908112-37-8 | 98.4% | 410.51 g/mol | C23H30N4O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14887710MG
Encompass_Preferred
AT-533 is a small-molecule inhibitor of Hsp90 with reported antiviral activity against herpes simplex virus (HSV). It suppresses tumor growth and angiogenesis by blocking the HIF-1α/VEGF/VEGFR-2 signaling pathway and inhibits downstream signaling including Akt/mTOR/p70S6K, Erk1/2, and FAK, making it useful as a research reagent in cell biology and preclinical studies.
  • Potent Hsp90 and HSV inhibitory activity.
  • Blocks HIF-1α/VEGF/VEGFR-2 signaling to suppress tumor growth and angiogenesis.
  • Inhibits downstream Akt/mTOR/p70S6K, Erk1/2, and FAK pathways.
  • Reduces tube formation, migration, and invasion in endothelial cell assays.
  • High purity suitable for research applications.
  • Stable as powder under recommended cold storage.
Catalog No. 50-002-85655
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.