Learn More
Medchemexpress LLC AT-9010 tetrasodium | 1621884-18-1 | 99.7% | 627.13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY139165A1MG
Encompass_Preferred
AT-9010 tetrasodium, a triphosphate active metabolite of AT-527, is a potent inhibitor of NiRAN (a function essential for viral replication). It can inhibit SARS-CoV-2 replication.
  • Potent inhibitor of NiRAN
  • Inhibits SARS-CoV-2 replication
Catalog No. 50-003-50752